Tisk…

Anti-MARCH4 Rabbit Polyclonal Antibody (Cy7®)

Dodavatel: Bioss
BOSSBS-9338R-CY7EA 24190 CZK
BOSSBS-9338R-CY7
Anti-MARCH4 Rabbit Polyclonal Antibody (Cy7®)
Protilátky
MARCH4, an E3 ubiquitin protein ligase, may mediate ubiquitination of MHC I and CD4, and promote their subsequent endocytosis and sorting to lysosomes via multivesicular bodies. E3 ubiquitin protein ligases accept ubiquitin from an E2 ubiquitin conjugating enzyme in the form of a thioester and then directly transfer the ubiquitin to targeted substrates.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MARCH4
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR