Tisk…

Anti-PSMA3 Rabbit Polyclonal Antibody (Cy5®)

Dodavatel: Bioss
BOSSBS-9352R-CY5EA 24190 CZK
BOSSBS-9352R-CY5
Anti-PSMA3 Rabbit Polyclonal Antibody (Cy5®)
Protilátky
The proteasome is a multicatalytic proteinase complex which is characterized by its ability to cleave peptides with Arg, Phe, Tyr, Leu, and Glu adjacent to the leaving group at neutral or slightly basic pH. The proteasome has an ATP-dependent proteolytic activity. Binds to the C-terminus of CDKN1A and thereby mediates its degradation. Negatively regulates the membrane trafficking of the cell-surface thromboxane A2 receptor (TBXA2R) isoform 2.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PSMA3
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR