Multitest slides, MP Biomedicals
Specifications
- Catalog No:
- ICNA096041205
- ICNA096041505
- ICNA096041805
- Colour:
- Blue
- Blue
- Blue
- W×D×H:
- 25×75×1 mm
- 25×75×1 mm
- 25×75×1 mm
Specifications
Product Information
- Catalog #Supplier #Growth areaNo. of wellsMaterialPkAvailabilityPrice
- Multitest slidesMultitest slidesICNA0960418050960418050,4 cm²10Glass1001 * 100 KS3 930,00 Kč
Specifications
ColourBlueEnvironmentally PreferableW×D×H25×75×1 mm - Multitest slidesMultitest slidesICNA0960412050960412050,4 cm²12Glass1001 * 100 KS4 230,00 Kč
Specifications
ColourBlueEnvironmentally PreferableW×D×H25×75×1 mm - Multitest slidesMultitest slidesICNA0960415050960415050,4 cm²15Glass1001 * 100 KS4 140,00 Kč
Specifications
ColourBlueEnvironmentally PreferableW×D×H25×75×1 mm