Tisk…
Eyewash solution, pH Neutral
Dodavatel: Plum Safety
118-1706EA 904 CZK
PLUM4746
Eyewash solution, pH Neutral
Bottle containing 4,9% sterile phosphate buffer solution for neutralising acids and alkali that come into contact with the eye.
  • Bottle equipped with a spray nozzle to ensure that the liquid can be distributed rapidly and evenly
  • User friendly design allows the bottles to be quickly opened with a single hand
  • Ergonomic eye-cups for effective flow
  • Suitable as refill for Plum station
Rinsing time: ~2 minutes
Shelf life: 3 years