Tisk…
Safety spectacles, Contour Metal
Dodavatel: Bollé Safety
Contour Metal
111-0388EA 633 CZK
111-0388 111-1602
Safety spectacles, Contour Metal
Brýle
Dual lens glasses with metal frame.
  • 160% flex temples
  • Non slip bridge
  • Increased upper protection
Weight: 38 g
Certifikace: EN 166, EN 170 (clear version), EN 172 (orange version)
Informace o dodávce: Supplied with microfibre pouch.