Tisk…

ABT-263 ≥98%

Dodavatel: BioVision

BIOV2467-25EA 19590 CZK
BIOV2467-25 BIOV2467-5
ABT-263 ≥98%
ABT-263

ABT-263 is an inhibitor of antiapoptotic Bcl-2 proteins.


  • DMSO (>50 mg/ml)


These inhibitors selectively binds to apoptosis suppressor proteins Bcl-2, Bcl-XL, and Bcl-w and prevents their binding to the apoptotic effectors Bax and Bak proteins, which may trigger apoptosis in tumor cells overexpressing Bcl-2, Bcl-XL, and Bcl-w.

Vzorec: C₄₅H₅₅ClF₃N₅O₆S₃
CAS číslo: 923564-51-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR