Tisk…

Cortisol (human/mouse/rat) ELISA kit

Dodavatel: BioVision
BIOVK7430-100EA 15730 CZK
BIOVK7430-100
Cortisol (human/mouse/rat) ELISA kit
Cortisol ELISA Kit is an in vitro assay for the quantitative measurement of cortisol in serum and plasma.

  • Detection method: Absorbance (450 nm)
  • Reactivity: Human, mouse, rat

Cortisol is a steroid hormone released from the adrenal cortex in response to the hormone ACTH. Cortisol acts through specific intracellular receptors and has effects on numerous physiologic systems, including immune system, glucose-counter regulation, vascular tone, substrate utilisation and bone metabolism.

Informace o dodávce: Kit includes 96-well plate coated with cortisol antibody, assay diluent, wash buffer (10X), standard diluent, cortisol standard, cortisol HRP conjugate (2000X), TMB susbstrate, stop solution and plate sealer.
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR