Tisk…
Polarising microscope
Dodavatel: Dino-Lite
IDCPAMK4112ZT-D15EA 17150 CZK
IDCPAMK4112ZT-D15
Polarising microscope
Designed for mobile applications, this digital microscope combines a polarisation filter with a portable recorder.
  • Anti-reflection polariser
  • Direct TV PAL RCA connection
  • Switchable white LED illumination
Certifikace: FCC, ROHS.
Informace o dodávce: Supplied with a USB cable, AC 110/240 V power adapter, rechargeable battery, mounting bracket for recorder, watertight sturdy case and a user manual.