Tisk…

[6,6]-Phenyl-C61-butyric acid methyl ester ≥99.9%, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL684457-100MGEA 43940 CZK
SIAL684457-100MG
[6,6]-Phenyl-C61-butyric acid methyl ester ≥99.9%, Sigma-Aldrich®
[6,6]-Phenyl-C61-butyric acid methyl ester

Soluble n-channel organic semiconductor. For use as an n-type layer in plastic electronics, especially bulk heterojunction OFETs and photovoltaic cells (PVs)


Synonyms: 1-[3-(Methoxycarbonyl)propyl]-1-phenyl-[6.6]C61, 3′H-Cyclopropa[1,9] [5,6]fullerene-C60-Ih-3′-butanoic acid 3′-phenyl methyl ester.

Vzorec: C₇₂H₁₄O₂
MW: 910,90 g/mol
MDL Number: MFCD07784544
CAS číslo: 160848-22-6

Objednat

Výsledky specifikačního testu

Assay 99,9%
Solubility Soluble in chlorobenzene, toluene and other organic solvents
Orbital energy (HOMO) 6,1 eV
Orbital energy (LUMO) 3,7 eV

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR