Tisk…
Sulphide indole motility (SIM) medium, Millipore®
Dodavatel: Merck
SIAL85438-100GEA 0 CZK
SIAL85438-100G
Sulphide indole motility (SIM) medium, Millipore®
Média mikrobiologická Mikrobiologická média dehydratovaná
Sulphide indole motility (SIM) medium is a test agar for Enterobacteriaceae, for the detection of motility and the formation of sulphide and indole.