Tisk…
Microlitre vials
548-2361EA 22830 CZK
VWRI548-2361
Microlitre vials
Microlitre vials with round foot 'Champagne vials' are designed as an alternative to fused insert and/or Total Recovery vials. These vials are made from clear or amber glass, 1st hydrolytic class.
  • Designed for use with nearly every autosampler
  • Self-standing
This vial design offers, on the one hand, a total max. volume of 1,1 ml by offering a very small residual volume of only 4 µl. This offers a wide range of common sample volumes to be used without changing the vial.