Tisk…
Indium standard solution
Dodavatel: LGC Standards
PROONIST-3124AEA 33370 CZK
PROONIST-3124A
Indium standard solution
Standardy Standardy kationické Standardy - indium
This reference standard is intended for use as a primary calibration standard for the quantitative determination of indium.
A unit consists of five 10 ml sealed borosilicate glass ampoules of solution prepared gravimetrically to contain a known mass fraction of indium.
Certifikace: Traceable to NIST and SRM numbers.