Tisk…

2-(Methacryloyloxy)ethyl-2-(trimethylammonio)ethyl phosphate ≥96.5% (by HPLC) stabilizovaný

Dodavatel: Merck


Stabilization: Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ) <100 ppm

SIAL730114-5GEA 5960 CZK
SIAL730114-5G
2-(Methacryloyloxy)ethyl-2-(trimethylammonio)ethyl phosphate ≥96.5% (by HPLC) stabilizovaný
2-(Methacryloyloxy)ethyl-2-(trimethylammonio)ethyl phosphate

Methacryloyloxyethyl phosphorylcholine (MPC), containing a phosphorylcholine group in the side chain is a most suitable monomer to mimic the phospholipid polar groups contained with cell membranes. MPC block copolymers are biocompatible in nature; it shows desirable interaction with living tissues. It is hydroscopic in nature.

Vzorec: C₁₁H₂₂NO₆P
MW: 295,27 g/mol
Bod tání: 143…148 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11112180
CAS číslo: 67881-98-5

Objednat

Výsledky specifikačního testu

Purity (HPLC) 96,5%
Colour White to light yellow and faint brown to light brown
Form Powder or Crystals
Carbon 43,2 - 46,3%
Nitrogen 4,4 - 4,9%
Inhibitor (MEHQ (Hydroquinone monomethyl ether)) <100 ppm
Infrared spectrum Conforms to structure
storage temperature 2 to 8 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR