Tisk…
Avantor® ACE® HILIC-A, HPLC Columns, Microbore
Dodavatel: Advanced Chromatography Technologies
ACE®
HILA-5-05005EA 29430 CZK
HILA-5-05005 HILA-3-15005 HILA-5-03005 HILA-3-05005S HILA-3-0501 HILA-5-05005S HILA-3-05005 HILA-5-15005 HILA-3-75005 HILA-5-25005 HILA-3-1001 HILA-3-12005 HILA-3-10005 HILA-3-03005S HILA-3-1501 HILA-5-03005S HILA-3-75005S HILA-5-10005 HILA-5-12005 HILA-5-75005 HILA-3-12005S HILA-5-75005S HILA-5-25005S HILA-5-10005S HILA-3-10005S HILA-5-12005S HILA-3-15005S HILA-3-03005 HILA-5-15005S
Avantor® ACE® HILIC-A, HPLC Columns, Microbore
Kolony chromatografické
Avantor® ACE® HILIC-A columns have been designed specifically for HILIC applications and are especially useful for the separation of polar analytes. These columns are part of a suit of 3 proprietary HILIC columns (HILIC-A, HILIC-B and HILIC-N) developed to enable chromatographers to achieve reproducible and successful HILIC separations using a systematic approach to method development.
  • Proprietary SIL (silica) acidic character HILIC phase
  • Compatible with all manufacturers’ HPLC/UHPLC systems
  • Excellent column performance, inertness and robustness
  • Simplified and structured approach to successful HILIC separations
  • Available in a range of particle sizes and column dimensions including UHPLC/HPLC rated hardware formats
Informace pro objednávání: HILIC method development kits, HILIC method development guide and flowchart are available on request.