Tisk…

Anti-ATG4B Rabbit Polyclonal Antibody

Dodavatel: ProSci
PRSI48-951EA 17450 CZK
PRSI48-951
Anti-ATG4B Rabbit Polyclonal Antibody
Protilátky
ATG4B antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 5 μg/mL, and immunofluorescence starting at 10 μg/mL.

Protein G Column

PBS, 0.2% gelatin, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ATG4B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chicken, Xenopus, Human, Mouse, Pufferfish, Rat, Zebrafish
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR