Tisk…

Anti-FOXO3 Rabbit Polyclonal Antibody

Dodavatel: ProSci
PRSI48-953EA 17280 CZK
PRSI48-953
Anti-FOXO3 Rabbit Polyclonal Antibody
Protilátky
FOXO3 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 10 μg/mL.

PBS, 0.05% BSA, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: FOXO3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Chicken, Dog, Xenopus, Human, Monkey, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR