Tisk…

Anti-ACHE Rabbit Polyclonal Antibody

Dodavatel: ProSci
PRSI48-958EA 17450 CZK
PRSI48-958
Anti-ACHE Rabbit Polyclonal Antibody
Protilátky
Acetylcholinesterase is involved in termination of signal transmission at neuromuscular junctions. Differences in assembly and localization of the multiple synaptic forms of acetylcholinesterase are thought to arise through posttranscriptional events. The Yt erythrocyte blood group antigen system resides on the acetylcholinesterase molecule.

Acetylcholinesterase antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 5 μg/mL, and immunofluorescence starting at 10 μg/mL.

PBS, 0.01% sodium azide, 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ACHE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR