Nechat relaci otevřenou?
Ukončení za 
Relace skončila
Vaše relace vypršela. Pro vaši bezpečnost jsme vás odhlásili.
Chcete se opět přihlásit?

Anti-DCK Rabbit Polyclonal Antibody

Dodavatel: ProSci Inc.
De oxycytidine kinase (DCK) is required for the phosphorylation of several deoxyribonucleosides and their nucleoside analogs. Deficiency of DCK is associated with resistance to antiviral and anticancer chemotherapeutic agents. Conversely, increased deoxycytidine kinase activity is associated with increased activation of these compounds to cytotoxic nucleoside triphosphate derivatives. DCK is clinically important because of its relationship to drug resistance and sensitivity.

Deoxycytidine Kinase antibody can be used in Western Blot starting at 0.5 - 2 μg/mL, and immunohistochemistry starting at 10 μg/mL.

PBS, 0.01% sodium azide, 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: DCK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human




 

OBJEDNÁVKA

Rozbalit vše / Zavřít vše
Rozměry protilátky Bal. VWR katalogové číslo Množství
50 µg 50 µG PRSI48-964
Generovat čárové kódy Formát popisku čárového kódu : 
Vygenerovat PDF katalog

Produktová specifikace

Symbol antigenu DCK
Název antigenu Deoxycytidine Kinase
Reaktivita Human
Spojení Unconjugated
Klonovalita Polyclonal
Hostitel Rabbit
Western blot Yes
ELISA Yes
Imunochemie Yes
Synonyma antigenu Deoxycytidine kinase, dCK
Imunogen Deoxycytidine Kinase antibody was raised against a synthetic peptide corresponding to N-Terminal residues of Deoxycytidine Kinase (Human).
Forma Liquid
Testované aplikace Immunoaffinity Chromatography
Enter Details

Select the article for which you are looking for Certificate

Doplňte Lot číslo. Toto pole je povinné. Jestli číslo neznáte, napíšte "x".

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Nebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Popis Katalogové číslo Availability Jednotka Vaše cena Price Per Množství