Tisk…
Avantor® ACE® HILIC-N, HPLC Columns, Microbore
Dodavatel: Advanced Chromatography Technologies
ACE®
HILN-5-15005SEA 29430 CZK
HILN-5-15005S HILN-5-10005S HILN-3-75005 HILN-3-1001 HILN-3-15005S HILN-5-15005 HILN-5-25005 HILN-5-75005 HILN-3-10005 HILN-3-1501 HILN-3-12005 HILN-5-03005S HILN-3-03005S HILN-3-12005S HILN-5-25005S HILN-5-12005S HILN-3-05005 HILN-3-03005 HILN-3-05005S HILN-3-0501 HILN-5-05005S HILN-5-03005 HILN-3-15005 HILN-5-10005 HILN-5-05005 HILN-3-10005S HILN-5-12005 HILN-3-75005S HILN-5-75005S
Avantor® ACE® HILIC-N, HPLC Columns, Microbore
Kolony chromatografické
Avantor® ACE® HILIC-N columns have been designed specifically for HILIC applications and are especially useful for the separation of polar analytes. These columns are part of a suit of 3 proprietary HILIC columns (HILIC-A, HILIC-B and HILIC-N) developed to enable chromatographers to achieve reproducible and successful HILIC separations using a systematic approach to method development.
  • Proprietary polyhydroxy neutral character HILIC phase
  • Compatible with all manufacturers’ HPLC/UHPLC systems
  • Excellent column performance, inertness and robustness
  • Simplified and structured approach to successful HILIC separations
  • Available in a range of particle sizes and column dimensions including UHPLC/HPLC rated hardware formats
Informace pro objednávání: HILIC method development kits, HILIC method development guide and flowchart are available on request.