About this item
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Product Information
- Catalog #Supplier #ElementSymbolConcentrationMatrixPkAvailabilityPrice
- PROOVHG-LLIN-100VHG-LLIN-100LithiumLi10 mg/L2% HNO₃100 mLLog in for availability1 * 100 mL1 810,00 KčSupplier Article Number: VHG-LLIN-100Local Article Number: PROOVHG-LLIN-100
- PROOVHG-PLIN-100VHG-PLIN-100LithiumLi1000 mg/L5% HNO₃100 mLLog in for availability1 * 100 mL1 640,00 KčSupplier Article Number: VHG-PLIN-100Local Article Number: PROOVHG-PLIN-100
- PROOVHG-PLIN-500VHG-PLIN-500LithiumLi1000 mg/L5% HNO₃500 mLLog in for availability1 * 500 mL3 610,00 KčSupplier Article Number: VHG-PLIN-500Local Article Number: PROOVHG-PLIN-500
- PROOVHG-TLIN-100VHG-TLIN-100LithiumLi10000 mg/L5% HNO₃100 mLLog in for availability1 * 100 mL3 050,00 KčSupplier Article Number: VHG-TLIN-100Local Article Number: PROOVHG-TLIN-100
- PROOVHG-TLIN-500VHG-TLIN-500LithiumLi10000 mg/L5% HNO₃500 mLLog in for availability1 * 500 mL6 660,00 KčSupplier Article Number: VHG-TLIN-500Local Article Number: PROOVHG-TLIN-500