Tisk…
AC sensor
Dodavatel: Brady
BRDY065269EA 1270 CZK
BRDY065269
AC sensor
AC sensor was selected by OSHA electrical equipment commitee as the only instrument of its type for use by OSHA compliance officers to identify and cite electrical hazards of 50 to 1000 V AC.
  • Simple to use and light weight
Colour: White on Red.
Certifikace: CAT IV rated and UL Listed.
Informace o dodávce: Includes 2 AAA battery (1,5 V).