About this item
CD19+ B cells are isolated from mononuclear cells by positive or negative selection using immunomagnetic cell separation procedures. Peripheral blood cells are collected from normal human volunteers by direct collection or by using the COBE® Spectra or Spectra Optia® Apheresis System where cell-rich leukapheresis product is extracted using continuous flow centrifugal technology directly into the collection bag.
- Purity >90% by flow cytometry
- Test negative for Hepatitis B and C, HIV, HTLV, HCV/HBV, WNV, Trypanasoma cruzi, and Syphilis
- Cryopreserved in CryoStor® CS10 media
CD19+ B cells play an essential role in the adaptive immune response. CD19+ B cells are responsible for the recognition and clearance of antigens in the system and, in the process, creating cells with the purpose of immunosurveillance. The wide range and sensitivity of the B cell to antigens lies in the process in which its receptor, the B cell receptor, develops.
Storage: Either prepare cells for long term storage in liquid nitrogen vapour phase or thaw for use. Storage in liquid phase nitrogen is NOT recommended. Short term storage of cells (less than 2 weeks) at –80°C is acceptable, but should be minimised to ensure maximum stability. Once thawed, samples must be used immediately.
Product Information
- Catalog #Supplier #Cell FormatPkAvailabilityPrice
- Human CD19+ B cells from peripheral blood, 5×106 cellsHuman CD19+ B cells from peripheral blood, 5×106 cellsPB19C-1PB19C-1Cryopreserved5 MouLog in for availability1 * 5 MouSupplier Article Number: PB19C-1Local Article Number: HCMSPB19C-1
- Human CD19+ B cells from peripheral blood, 10×106 cellsHuman CD19+ B cells from peripheral blood, 10×106 cellsPB19C-2PB19C-2Cryopreserved10 MouLog in for availability1 * 10 MouSupplier Article Number: PB19C-2Local Article Number: HCMSPB19C-2
- Human CD19+ B cells from peripheral blood, 25×106 cellsHuman CD19+ B cells from peripheral blood, 25×106 cellsPB19C-3PB19C-3Cryopreserved25 MouLog in for availability1 * 25 MouSupplier Article Number: PB19C-3Local Article Number: HCMSPB19C-3
- Human CD19+ B cells from peripheral blood, negative selection, 10×106 cellsHuman CD19+ B cells from peripheral blood, negative selection, 10×106 cellsPB19NC-1PB19NC-1Cryopreserved10 MouLog in for availability1 * 10 Mou31 430,00 KčSupplier Article Number: PB19NC-1Local Article Number: HCMSPB19NC-1