Tisk…
Human plasma, lung cancer (LC)
Dodavatel: Charles River Laboratories Cell Solutions
HCMSPB006C-1-LCEA 0 CZK
HCMSPB006C-1-LC
Human plasma, lung cancer (LC)
Human peripheral blood plasma used for researching the causes, diagnosis, supportive care and treatment of lung cancer (LC). Collected from IRB-consented donors diagnosed with LC.
Patient samples can be used by researchers to investigate biomarker and diagnostic development, pre-clinical research validation, and drug toxicity and efficacy.
No pre-screening is done on fresh products, but donors are tested on day of collection for Hepatitis B, Hepatitis C, and HIV 1/2.
Storage: Store at −80 °C.
Upozornení: Intended for research use only and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.