Tisk…
Whole blood, chronic myeloid leukemia (CML)
HCMSPB000F-2-CMLEA 0 CZK
HCMSPB000F-2-CML
Whole blood, chronic myeloid leukemia (CML)
Whole Blood used for researching the causes, diagnosis, supportive care and treatment of chronic myeloid leukemia (CML). Collected from IRB-consented donors diagnosed with chronic myeloid leukemia (CML).
No pre-screening is done on fresh products, but donors are tested on day of collection for Hepatitis B, Hepatitis C, and HIV 1/2, HTLV-I/II, HIV-1/HCV/HBV Nucleic Acid, WNV Nucleic Acid, Trypanasoma cruzi and Syphilis.
Storage: Use immediately upon receipt.
Upozornení: Intended for only research use and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.