Tisk…
Bottles, wide neck, with standard ground stoppers
Dodavatel: Hecht
HECH41287018EA 233 CZK
HECH41287018 HECH41285006
Bottles, wide neck, with standard ground stoppers
Láhve Láhve skleněné
Bottles are clear or brown with standard ground joint and glass stopper.
  • Good chemical and physical properties
  • Available with label and fused inscription