Tisk…

Hydrogen peroxide assay kit

Dodavatel: Abnova
ABNOKA0801EA 15540 CZK
ABNOKA0801
Hydrogen peroxide assay kit
Test Enzymatická stanovení
Hydrogen Peroxide Assay Kit is used for measuring H2O2 in biological samples.

  • Detection method: Colorimetric, fluorometric
  • Reactivity: Mammals
  • Suitable samples include biological fluid, cell culture supernatant, plasma, serum, tissue lysate, urine

Informace o dodávce: Kit contains H2O2 assay buffer (25 ml), OxiRed probe, DMSO (0,4 ml), HRP, H2O2 Standard 0,88 M (0,1 ml) .
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR