Tisk…
Vacuum controller, VC 10
Dodavatel: IKA
IKAA20005132EA 73840 CZK
IKAA20005132
Vacuum controller, VC 10
Čerpadla Kontroléry vakua
Universal vacuum controller for a two-point calibration of multiple vacuum sources. It comes with a chemo resistant ceramic pressure sensor.
  • High-resolution TFT display ensures intuitive operation and navigation
  • ventilation valve works even during a power outage and aerates the vacuum chamber
In addition to the manual mode, up to 10 programs can be saved, with upto 10 pressure/time sequences, even allowing for saving the last experiment run. It is possible to manage the experiment via labworldsoft (RS 232 interface) and to connect an external PT 1000 temperature sensor to measure the temperature of all applications.