Tisk…
Microinjectors, FemtoJet® 4i
Dodavatel: Eppendorf
FemtoJet®
EPPE5252000.013EA 0 CZK
EPPE5252000.013
Microinjectors, FemtoJet® 4i
Mikroinjektory Mikroinjektory Programovatelné
The FemtoJet® 4i is a programmable microinjector with integrated pressure supply suitable for injecting small to intermediate volumes (up to 100 pl).
  • Very simple and clearly arranged operation for routine tasks
  • Injection volume ranges from femtoliters to microliters
  • Inject and Clean function can be triggered right on the device using the included mouse or foot control
  • Programmable injection guarantees high reproducibility
  • Connection to TransferMan® 4r, InjectMan® 4 or InjectMan® NI 2 enables semi-automatic microinjection
  • Can be combined with all conventional micromanipulation systems
  • Independent pressure supply via built-in compressor
Clean function: Maximum rinsing pressure of 6000 hPa
Injection time: 0,01 - 99,99 s; can be set in increments of 0,01 s