Tisk…
Sleeve protectors, heat resistant, HyFlex®
Dodavatel: Ansell
ANSE70-415EA 4030 CZK
ANSE70-415 ANSE70-414 ANSE70-419 ANSE70-416 ANSE70-422 ANSE70-410
Sleeve protectors, heat resistant, HyFlex®
These sleeve protectors are made from high-tech 100% Kevlar® para-aramid short fibers for an exceptional cut resistance and comfort.
  • Thumb slot ensures that the sleeve stays in place
  • Double layered fabric, silicone free and latex free
  • Protect the arm against heat (up to 100 °C)
  • Fine stretchable fabric provides excellent comfort
  • Not for use with liquids or pointed objects
Washing temperature is 40 °C (104 °F). Bar tack sleeve to hold the sleeve in position. Unique 'stay-up' band ensures sleeves remaining in place.
Certifikace: EN 388 (134X), EN 407 (X1XXXX).