Tisk…
BIOV2497
BIOV2497-250EA 3050 CZK
BIOV2497-250 BIOV2497-5G BIOV2497-1G
BIOV2497
Amphotericin B is a polyene antifungal antibiotic from Streptomyces which targets a wide range of susceptible fungi including Histoplasma, Coccidioides, and Cryptococcus species.
  • Appearence: Yellow or orange powder
  • Solubility: DMF (~2 – 4 mg/ml) or DMSO (~30 – 40 mg/ml)
Amphotericin B targets ergosterol in the fungal cell membrane and increases permeability. This alteration of the cell membrane is toxic to the fungus and leads to death. Can be used as a selective agent in several types of isolation media.