Tisk…
Plunger cans, premium line
Dodavatel: asecos
MSGA33533EA 3010 CZK
MSGA33533 MSGA33532
Plunger cans, premium line
Nádoby bezpečnostní
These plunger cans are either designed with inside tin coating with high-quality powder coating on the outside or made of PE.
  • Ideal for economical wetting of cleaning cloths
  • Pumping station made of brass
  • Includes with container identification method
  • High stability
These are mainly involved in efficient removal of the cleaning fluids by reducing the leakage of fumes. Designed with spring-mounted potion plate with flame arrester.
Compliance with FM and TÜV approved.