Tisk…
Protective netting, Poly-net®
Dodavatel: Merck
SIALZ183709-5MEA 0 CZK
SIALZ183709-5M SIALZ183687-5M
Protective netting, Poly-net®
Návleky ochranné
The Poly-net® protective netting is made from PE net sleeve which stretches around glassware to prevent scratching, chipping or breaking.
  • Netting captures flying glass if container shatters
  • Easily cut to any length
  • Color-coded for size identification
Ideal for use with funnels, flasks or condensers.
Thickness: 0,07 to 1,2 mm