Tisk…
Pipette Stand 2 and Charger Stand 2
Dodavatel: Eppendorf
EPPE3116000.040EA 7840 CZK
EPPE3116000.040 EPPE3116000.031 EPPE3116000.058
Pipette Stand 2 and Charger Stand 2
Stojany na pipety
These stands for single instruments have a robust design and securely hold one manual or electronic device. With the Charger Stand 2, your Eppendorf Xplorer®, Eppendorf Xplorer plus or Multipette® E3/E3x is always fully charged and ready for use. The Pipette Stand 2 carries one Eppendorf Multipette M4.
  • Large rubber feet protect against liquids spilled on bench top
  • Small footprint with a diameter of 18,5 cm
  • Fast interchangeable pipette holders and charger shells, no tool required
  • Pictograms clearly label pipette holders and charger shells for easy assignment in the lab
  • Cord wrap functionality for unused power cord (Charger)
  • Magnetic power connector for fast and easy connection
The Pipette Stand 2 is also perfect for one single or multi-channel Eppendorf Research® plus or Eppendorf Reference® 2 pipette when used with the corresponding pipette holder.