Silikagel 40% in water, colloidal dispersion
12727.A1EA
1090
CZK
12727.A1
12727.A7
Silikagel 40% in water, colloidal dispersion
Silicon dioxide
Vzorec:
SiO₂ MW: 60,08 g/mol Hustota: 1,30 Storage Temperature: Ambient |
MDL Number:
MFCD00011232 CAS číslo: 7631-86-9 EINECS: 231-545-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...