Bond tack cloth
CONTBS20CB-12
: CONTEC
- Packed:100/bag, 5 bags/case
- Description:Tack cloth
- L×W:229×305 mm
- Pk:500
These polyester cloths are ideal for the surface preparation prior to painting. Also suitable for particle removal in printed circuit board manufacturing and for eliminating solvents in some applications.
- Custom blended tack formula for optimal adhesive texture, transfer resistance, heat resistance and anti-static performance
- Sealed edge technology minimises particles and fibres
- "Napped" surface easily removes dirt particles
- Water-based tack is applied at high heat under pressure