Indium, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
SIAL326615-50GEA
15470
CZK
SIAL326615-50G
Indium, Sigma-Aldrich®
Indium
Vzorec:
In MW: 114,82 g/mol Bod varu: 2000 °C (1013 hPa) Bod tání: 156 °C Hustota: 7,3 g/cm³ (20 °C) |
MDL Number:
MFCD00134048 CAS číslo: 7440-74-6 EINECS: 231-180-0 Merck Index: 13,04971 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...