Indium, powder, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
SIAL203432-5GEA
11380
CZK
SIAL203432-5G
SIAL203432-25G
Indium, powder, Sigma-Aldrich®
Indium
Vzorec:
In MW: 114,82 g/mol Hustota: 7,31 g/cm³ (20 °C) |
MDL Number:
MFCD00134048 CAS číslo: 7440-74-6 EINECS: 231-180-0 UN: 3089 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...