Tisk…
Beaker cups, PP, disposable
Dodavatel: Semadeni
SEMA8611EA 236 CZK
SEMA8611
Beaker cups, PP, disposable
For use as drinking cups, in laboratories and as sample cups.
  • Very stable
  • With graduation
  • Good chemical and temperature resistance
Capacity: 30 ml