cín ≥99.85% (metals basis), rods, Ø 6.35 cm (2.5 in)
12601.36EA
6080
CZK
12601.36
cín ≥99.85% (metals basis), rods, Ø 6.35 cm (2.5 in)
cín
Vzorec:
Sn MW: 118,71 g/mol Bod varu: 2270 °C (1013 hPa) Bod tání: 232 °C Hustota: 7,31 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00133862 CAS číslo: 7440-31-5 EINECS: 231-141-8 Merck Index: 12,09587 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...