Zlato, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
SIAL349305-1.5GEA
17060
CZK
SIAL349305-1.5G
Zlato, Sigma-Aldrich®
Zlato
Vzorec:
Au MW: 196,97 g/mol Bod varu: 2800 °C (1013 hPa) Bod tání: 1063 °C Hustota: 19,3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS číslo: 7440-57-5 EINECS: 231-165-9 Merck Index: 12,04526 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...