Tisk…

Zlato, powder, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL255734-1GEA 10150 CZK
SIAL255734-1G
Zlato, powder, Sigma-Aldrich®
Zlato
Vzorec: Au
MW: 196,97 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS číslo: 7440-57-5
EINECS: 231-165-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR