Tisk…
Zirconium ≥99.5% (metals basis excluding Hf), slug (Hf ∼ 3%), Length: 12.7 mm (0.50 in), Ø 6.35 mm (0.25 in)
Dodavatel: Thermo Scientific
42508.14EA 1340 CZK
42508.14 42508.22
Zirconium ≥99.5% (metals basis excluding Hf), slug (Hf ∼ 3%), Length: 12.7 mm (0.50 in), Ø 6.35 mm (0.25 in)
Zirconium
Vzorec: Zr
MW: 91,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011303
CAS číslo: 7440-67-7
EINECS: 231-176-9