|
|
OBJEDNÁVKA
Produktová specifikace
Čí. CAS | 7439-98-7 | ||
Purity | 99.8% | ||
Fyzická forma | ≈0.0014g/m: Temper: hard | ||
MDL čí. | MFCD00003465 |
![]() |
Zpracováváme váš požadavek… |
Chcete se opravdu vrátit? |
||
|
položek přidáno do vašeho nákupního košíku
|
||||||||
---|---|---|---|---|---|---|---|---|
|
With support from VWRCATALYST, your business can maintain a competitive edge.
Our services re-focus scientific time to help you accelerate productivity and drive innovation, from research through production.
Over 1,200 VWRCATALYST associates are working worldwide today at industry-leading pharmaceutical, biotech, healthcare, education, industrial, and high-tech production institutions.
Contact us at vwrcatalyst.eu@vwr.com.
Rozwiązania bioprocesowe VWR umożliwiają producentom biofarmaceutycznym wdrażanie technologii, które przyspieszają innowacje naukowe.
Dowiedz się więcej o naszych możliwościach i pobierz broszurę.
![]() |
|
|
Avantor is a leading global provider of integrated, tailored solutions for the life sciences and advanced technology industries. Strengthened by the recent acquisition of VWR, the Company is a trusted partner to customers and suppliers from discovery to delivery. Collectively, we set science in motion to create a better world.