Železo ≥99.99% (metals basis), foil, Thickness: 0.5 mm (0.02 in)
40495.FFEA
3990
CZK
40495.FF
40495.GH
40495.FI
Železo ≥99.99% (metals basis), foil, Thickness: 0.5 mm (0.02 in)
Železo
Vzorec:
Fe MW: 55,85 g/mol Bod varu: 3000 °C (1013 hPa) Bod tání: 1535 °C Hustota: 7,86 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010999 CAS číslo: 7439-89-6 EINECS: 231-096-4 Merck Index: 13,05109 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...