Tisk…
Aconitase assay kit
Dodavatel: Abnova
ABNOKA0876EA 27310 CZK
ABNOKA0876
Aconitase assay kit
Test Enzymatická stanovení
Aconitase Assay Kit is used for measuring aconitase activity.
  • Detection method: Colorimetric
  • Reactivity: Mammals
  • Storage temperature: 4 °C
Aconitase (EC 4.2.1.3) is an iron-sulphur protein containing an [Fe₄S₄]²⁺ cluster that catalyses the stereospecific isomerisation of citrate to isocitrate via cis-aconitate in the tricarboxylic acid cycle, a non-redox-active process. This assay kit is highly sensitive, simple, direct and HTS-ready colorimetric assay for measuring aconitase activity in biological samples. In the assay, citrate is converted by aconitase into isocitrate, which is further processed resulting in a product that converts a nearly colourless probe into an intensely coloured form (λ= 450 nm).
Informace o dodávce: Kit contains aconitase assay buffer (30 ml), enzyme mix (200 µl) and each one vial of substrate, developer, cysteine, (NH₄)Fe(SO₄)₂, isocitrate standard.
Upozornení: For research use only.