Tisk…

Paladium ≥99.9% (trace metals basis), powder <75 µm, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí


M-Clarity™ quality level = MQ100

SIAL464651-5GEA 23690 CZK
SIAL464651-5G
Paladium ≥99.9% (trace metals basis), powder <75 µm, Sigma-Aldrich®
Palladium
Vzorec: Pd
MW: 106,420 g/mol
Hustota: 12,02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS číslo: 7440-05-3
EINECS: 231-115-6
ADR: 4.1,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR