Platina, wire for ISA type R or S standard grade thermocouple, Ø 0.0508 mm (0.002 in)
Dodavatel: Thermo Scientific
14361.BUEA
9370
CZK
14361.BU
14361.BY
Platina, wire for ISA type R or S standard grade thermocouple, Ø 0.0508 mm (0.002 in)
Platina
Vzorec:
Pt MW: 195,08 g/mol Bod varu: 3827 °C (1013 hPa) Bod tání: 1769 °C Hustota: 21,45G/CC Storage Temperature: Ambient |
MDL Number:
MFCD00011179 CAS číslo: 7440-06-4 EINECS: 231-116-1 Merck Index: 12,07689 |