Tisk…

Colorimetric collagenase activity assay kit

Dodavatel: BioVision
BIOVK792-100EA 10490 CZK
BIOVK792-100
Colorimetric collagenase activity assay kit
Colorimetric Collagenase Activity Assay Kit provides a quick and easy way to determine activity of collagenase. The kit measures collagenase activity using a synthetic peptide (FALGPA) that mimics collagen’s structure. It is suitable for measuring activity of bacterial collagenases such as from Clostridium histolyticum type I-XI. In addition, it can also be used to screen/characterise collagenase inhibitors. The limit of detection for this assay is 0,02 mU collagenase.

  • Rapid and sensitive
  • Species reactivity: Bacterial
  • Detection method: Absorbance (345 nm)

Informace o dodávce: Kit includes collagenase assay buffer, collagenase (0,35 U/ml), collagenase substrate (FALGPA) and 1 M inhibitor (1,10-Phenanthroline).

Upozornení: For research use only.
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR