Tisk…
Chamber furnaces, N 7/H
Dodavatel: Nabertherm
NABEN01H0NCNAEA 91000 CZK
NABEN01H0NCNA NABEN01H0NBNA
Chamber furnaces, N 7/H
Benchtop chamber furnaces for annealing, hardening and brazing.
  • Three-sided heating from both sides and chamber floor
  • Heating elements on support tubes ensure free heat radiation and a long service life
  • Heating elemenets on the floor are protected by heat-resistant SiC plate
  • Low energy consumption due to multi-layer insulation
  • Parallel guided downward swinging door (protects user from heat radiation)
  • Gas spring dampers ensure easy door opening and closing
  • NTLog Basic for Nabertherm controller: Records process data with USB-flash drive
Even high-performance applications such as annealing of titanium in medical applications can be carried out without requiring complicated annealing systems.
Weight: 60 kg
Informace o dodávce: Supplied with base frame
Upozornení: Note: Maximum continuous operating temperature should be 50 °C below the max. temp. of the model shown in the table above.