Tisk…

Agar, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ300

SIALW201201-100GEA 0 CZK
SIALW201201-100G SIALW201201-1KG SIALW201201-SAMPLE
Agar, Sigma-Aldrich®
Agar
Bod tání: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00081288
CAS číslo: 9002-18-0
EINECS: 232-658-1
Merck Index: 13,00184
https://cz.vwr.com/stibo/web/std.lang.all/81/91/2298191.jpg

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR